ENA161209695 Enamine

6-chloro-1-ethyl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

  • MDL number MFCD00206329
  • Cas Number : 66176-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-36078

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36078
Availability details for
Quantity
  • Availability
  • Add to Cart