ENA161209704 Enamine

2-[2-(trifluoromethyl)phenoxy]acetic acid

  • MDL number MFCD06409075
  • Cas Number : 162922-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36088

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36088
Availability details for
Quantity
  • Availability
  • Add to Cart