ENA161209710 Enamine

2-methyl-2-pyrrolidin-1-ylpropanenitrile

  • MDL number MFCD10005350
  • Cas Number : 35666-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36094

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36094
Availability details for
Quantity
  • Availability
  • Add to Cart