ENA161209713 Enamine

N-(1-cyclohexanecarbonylpiperidin-4-ylidene)hydroxylamine

  • MDL number MFCD09950105
  • Cas Number : 1016888-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36097
Availability details for
Quantity
  • Availability
  • Add to Cart