ENA161209732 Enamine

4-methoxy-3-(2,2,2-trifluoroethoxy)aniline

  • MDL number MFCD09730834
  • Cas Number : 953744-38-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-36121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36121
Availability details for
Quantity
  • Availability
  • Add to Cart