ENA161209737 Enamine

1-(2,4-difluorophenyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD08444772
  • Cas Number : 923178-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36133

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36133
Availability details for
Quantity
  • Availability
  • Add to Cart