ENA161209739 Enamine

1-[2-(1H-pyrazol-1-yl)pyridin-3-yl]methanamine

  • MDL number MFCD09739780
  • Cas Number : 1052559-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36135
Availability details for
Quantity
  • Availability
  • Add to Cart