ENA161209753 Enamine

2-(4-methyl-2-phenylpiperazin-1-yl)ethan-1-amine

  • MDL number MFCD09810914
  • Cas Number : 1016715-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35768

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35768
Availability details for
Quantity
  • Availability
  • Add to Cart