ENA161209758 Enamine

1-(1-benzofuran-2-carbonyl)piperazine

  • MDL number MFCD08444706
  • Cas Number : 41717-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36172
Availability details for
Quantity
  • Availability
  • Add to Cart