ENA161209761 Enamine

4-[(4-methoxybenzoyl)amino]butanoic acid

  • MDL number MFCD00434431
  • Cas Number : 72432-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122...124
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36177

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36177
Availability details for
Quantity
  • Availability
  • Add to Cart