ENA161209781 Enamine

2-(4-aminopiperidin-1-yl)-N-methylacetamide

  • MDL number MFCD09811200
  • Cas Number : 882562-53-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36235
Availability details for
Quantity
  • Availability
  • Add to Cart