ENA161209782 Enamine

5-(aminomethyl)-N,N-dimethylpyridin-2-amine

  • MDL number MFCD08272104
  • Cas Number : 354824-17-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-35897

Safety Information

msds https://www.enaminestore.com/catalog/EN300-35897
Availability details for
Quantity
  • Availability
  • Add to Cart