ENA161209802 Enamine

2-(8-methyl-4-oxo-3,4-dihydroquinazolin-2-yl)acetonitrile

  • MDL number MFCD06642597
  • Cas Number : 477879-84-8
  • Molecular Weight : 199.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-35906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-35906
Availability details for
Quantity
  • Availability
  • Add to Cart