ENA161668947 Enamine

1-chloroisoquinoline-3-carboxylic acid

  • MDL number MFCD10686865
  • Cas Number : 1049606-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36217
Availability details for
Quantity
  • Availability
  • Add to Cart