ENA161668951 Enamine

1-[(3,5-dimethyl-1,2-oxazol-4-yl)methyl]piperazine dihydrochloride

  • MDL number MFCD10686874
  • Cas Number : 333992-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36259

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36259
Availability details for
Quantity
  • Availability
  • Add to Cart