ENA161668957 Enamine

3-methyl-1-(piperidine-1-carbonyl)-1H-imidazol-3-ium iodide

  • MDL number MFCD10686884
  • Cas Number : 548763-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data I-
Product Link https://www.enaminestore.com/catalog/EN300-36297

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36297
Availability details for
Quantity
  • Availability
  • Add to Cart