ENA161668959 Enamine

3-amino-N-[4-(hydrazinecarbonyl)phenyl]benzene-1-sulfonamide

  • MDL number MFCD10686886
  • Cas Number : 1049606-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-36311

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36311
Availability details for
Quantity
  • Availability
  • Add to Cart