ENA161668961 Enamine

3-(dimethylamino)propanethioamide hydrochloride

  • MDL number MFCD10686889
  • Cas Number : 24812-53-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36371
Availability details for
Quantity
  • Availability
  • Add to Cart