ENA161668962 Enamine

N-benzyl-2-[5-(chloromethyl)-1,2,4-oxadiazol-3-yl]acetamide

  • MDL number MFCD10686890
  • Cas Number : 1050886-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-36375

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36375
Availability details for
Quantity
  • Availability
  • Add to Cart