ENA161668963 Enamine

3-[(4-methylphenyl)amino]propanethioamide

  • MDL number MFCD10686891
  • Cas Number : 1049874-17-0
  • Molecular Weight : 194.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-36376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36376
Availability details for
Quantity
  • Availability
  • Add to Cart