ENA161668965 Enamine

sodium 2-{2-[(3-chlorophenyl)methyl]-1,3-thiazol-4-yl}acetate

  • MDL number MFCD10686893
  • Cas Number : 1171566-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36381
Availability details for
Quantity
  • Availability
  • Add to Cart