ENA161668969 Enamine

1-(2-amino-4-methylpyrimidin-5-yl)ethanone oxime

  • MDL number MFCD10686898
  • Cas Number : 1050886-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36392

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36392
Availability details for
Quantity
  • Availability
  • Add to Cart