ENA161668998 Enamine

N-[(3-aminophenyl)methyl]-N-methylacetamide

  • MDL number MFCD10686937
  • Cas Number : 1049874-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36704

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36704
Availability details for
Quantity
  • Availability
  • Add to Cart