ENA161669000 Enamine

N-[(4-fluorophenyl)methyl]-1-(N'-hydroxycarbamimidoyl)formamide

  • MDL number MFCD10686939
  • Cas Number : 1049607-08-0
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-36714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36714
Availability details for
Quantity
  • Availability
  • Add to Cart