ENA161669001 Enamine

2-chloropyridine-4-carbothioamide

  • MDL number MFCD10686940
  • Cas Number : 91447-89-1
  • Molecular Weight : 172.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-36717

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36717
Availability details for
Quantity
  • Availability
  • Add to Cart