ENA161669002 Enamine

5,6-dimethyl-3-oxo-2,3-dihydropyridazine-4-carboxamide

  • MDL number MFCD10686941
  • Cas Number : 1050886-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-36720

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36720
Availability details for
Quantity
  • Availability
  • Add to Cart