ENA161669005 Enamine

3-(1H-1,2,3,4-tetrazol-1-yl)thiophene-2-carboxylic acid

  • MDL number MFCD10686944
  • Cas Number : 1049874-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36731
Availability details for
Quantity
  • Availability
  • Add to Cart