ENA161669078 Enamine

methyl 2-chloro-5-(morpholine-4-sulfonyl)benzoate

  • MDL number MFCD01203055
  • Cas Number : 445224-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28649
Availability details for
Quantity
  • Availability
  • Add to Cart