ENA161669083 Enamine

2-chloro-1,3-thiazole-5-sulfonyl chloride

  • MDL number MFCD09742125
  • Cas Number : 88917-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-29708

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-29708
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart