ENA161669111 Enamine

8-fluoroquinoline-5-sulfonyl chloride

  • MDL number MFCD09702325
  • Cas Number : 1000933-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36180

Safety Information

signalword Warning
pcodes P264,P270,P301+P312,P330,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-36180
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart