ENA161669122 Enamine

1-(5-chloro-2-methoxybenzoyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD09812458
  • Cas Number : 1101860-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36220

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36220
Availability details for
Quantity
  • Availability
  • Add to Cart