ENA161669124 Enamine

8-iodo-1,3-dimethyl-3,9-dihydro-1H-purine-2,6-dione

  • MDL number MFCD00160128
  • Cas Number : 24255-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16742

Safety Information

msds https://www.enaminestore.com/catalog/EN300-16742
Availability details for
Quantity
  • Availability
  • Add to Cart