ENA161669133 Enamine

6-(isopropylamino)nicotinonitrile

  • MDL number MFCD09944214
  • Cas Number : 160017-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36247

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36247
Availability details for
Quantity
  • Availability
  • Add to Cart