ENA161669136 Enamine

1-(2,4-dimethoxybenzoyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD09807946
  • Cas Number : 1103891-72-0
  • Molecular Weight : 279.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-36250

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36250
Availability details for
Quantity
  • Availability
  • Add to Cart