ENA161669147 Enamine

5-ethylfuran-2-carboxylic acid

  • MDL number MFCD04378818
  • Cas Number : 56311-37-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90-92
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36265
Availability details for
Quantity
  • Availability
  • Add to Cart