ENA161669174 Enamine

1-(3-aminophenyl)piperidin-2-one

  • MDL number MFCD09743290
  • Cas Number : 69131-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36314

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36314
Availability details for
Quantity
  • Availability
  • Add to Cart