ENA161669179 Enamine

6-methoxy-3,4-dihydro-2H-1-benzopyran-3-carboxylic acid

  • MDL number MFCD04114628
  • Cas Number : 182570-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36319
Availability details for
Quantity
  • Availability
  • Add to Cart