ENA161669182 Enamine

5-(2-chlorophenyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD04327495
  • Cas Number : 1197631-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225...227
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36322

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36322
Availability details for
Quantity
  • Availability
  • Add to Cart