ENA161669200 Enamine

6,8-difluoro-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD09041806
  • Cas Number : 926218-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-36382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36382
Availability details for
Quantity
  • Availability
  • Add to Cart