ENA161669204 Enamine

2-(dimethylamino)pyridine-4-carbonitrile

  • MDL number MFCD09937878
  • Cas Number : 80882-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70-72
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-36388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36388
Availability details for
Quantity
  • Availability
  • Add to Cart