ENA161669218 Enamine

1-methyl-4-(piperidine-1-sulfonyl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD09052822
  • Cas Number : 937625-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36405

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36405
Availability details for
Quantity
  • Availability
  • Add to Cart