ENA161669220 Enamine

5-(piperidine-1-sulfonyl)furan-2-carboxylic acid

  • MDL number MFCD10686901
  • Cas Number : 933735-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36407

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36407
Availability details for
Quantity
  • Availability
  • Add to Cart