ENA161669247 Enamine

N-{4-[1-(hydroxyimino)ethyl]phenyl}propanamide

  • Cas Number : 1016878-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-36479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36479
Availability details for
Quantity
  • Availability
  • Add to Cart