ENA161669250 Enamine

1-(3,4-dichlorobenzoyl)piperidine-3-carboxylic acid

  • MDL number MFCD09050556
  • Cas Number : 926246-92-6
  • Molecular Weight : 302.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-36482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36482
Availability details for
Quantity
  • Availability
  • Add to Cart