ENA161669260 Enamine

3-(2-fluorobenzamido)-4-methylbenzoic acid

  • MDL number MFCD09049042
  • Cas Number : 926237-97-0
  • Molecular Weight : 273.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-36494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36494
Availability details for
Quantity
  • Availability
  • Add to Cart