ENA161669263 Enamine

4-(2-methylpropoxy)benzene-1-sulfonyl chloride

  • MDL number MFCD09804087
  • Cas Number : 1138-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36497
Availability details for
Quantity
  • Availability
  • Add to Cart