ENA161669270 Enamine

2-(4-aminophenyl)-N-[2-(dimethylamino)ethyl]acetamide

  • MDL number MFCD09044244
  • Cas Number : 331759-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36505
Availability details for
Quantity
  • Availability
  • Add to Cart