ENA161669283 Enamine

[(cyclopropylcarbonyl)amino]acetic acid

  • MDL number MFCD01724840
  • Cas Number : 64513-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36522

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36522
Availability details for
Quantity
  • Availability
  • Add to Cart