ENA161669284 Enamine

5-(2-chlorophenyl)-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD06005852
  • Cas Number : 334017-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36523

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36523
Availability details for
Quantity
  • Availability
  • Add to Cart