ENA161669294 Enamine

4-[(pyridin-4-yl)methoxy]benzaldehyde

  • MDL number MFCD08444878
  • Cas Number : 118001-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36537

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36537
Availability details for
Quantity
  • Availability
  • Add to Cart